Arsenic(III) chloride ≥99.999% (trace metals basis)
Supplier: BeanTown Chemical
145185-25G
145185-5G
BT145185-25GEA
130
USD
BT145185-25G
BT145185-5G
Arsenic(III) chloride ≥99.999% (trace metals basis)
Arsenic(III) chloride
CAS: 7784-34-1; EC No: 232-059-5; MDL No: MFCD00014165; RTECS: CG1750000
UN No: UN1560; Haz Class: 6.1; Packing Group: I
Liquid; Molecular Formula: AsCl3; MW: 181.28
Melting Point: -16°; Boiling Point: 130.2°
Density (g/mL): 2.15
Moisture Sensitive
- RTECS: CG1750000
- Moisture Sensitive
- Liquid
Formula:
AsCl₃ MW: 181.28 g/mol Boiling Pt: 130.2 °C Melting Pt: –16 °C Density: 2.15 g/mL |
MDL Number:
MFCD00014165 CAS Number: 7784-34-1 EINECS: 232-059-5 UN: 1560 ADR: 6.1,I |
Specification Test Results
Assay | 99.999% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...