3-Chloroisonicotinonitrile ≥95%
Supplier: BeanTown Chemical
145055-1G
145055-5G
BT145055-1GEA
799
USD
BT145055-1G
BT145055-5G
3-Chloroisonicotinonitrile ≥95%
3-Chloroisonicotinonitrile
CAS: 68325-15-5; MDL No: MFCD05663706
UN No: UN3439; Haz Class: 6.1; Packing Group: III
Solid; Molecular Formula: C6H3ClN2; MW: 138.55
Melting Point: 70-73°
- Solid
Formula:
C₆H₃ClN₂ Melting Pt: 70…73 °C |
MDL Number:
MFCD05663706 CAS Number: 68325-15-5 UN: 3439 ADR: 6.1,III |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...