Silver bromide ≥99.995%

Supplier: BeanTown Chemical

Synonyms: Argentous bromide

145030-100G 145030-25G 145030-5G
BT145030-100GEA 561 USD
BT145030-100G BT145030-25G BT145030-5G
Silver bromide ≥99.995%
Silver bromide

CAS: 7785-23-1; EC No: 232-076-8; MDL No: MFCD00003398 UN No: UN3077; Haz Class: 9; Packing Group: III Crystalline; Molecular Formula: AgBr; MW: 187.78 Melting Point: 432° Density (g/mL): 6.473 Light Sensitive


  • Light Sensitive
  • Crystalline

Formula: AgBr
MW: 187.77 g/mol
Boiling Pt: 1300
Melting Pt: 432
Density: 6.473 g/mL
MDL Number: MFCD00003398
CAS Number: 7785-23-1
EINECS: 232-076-8
ADR: 9 III
Merck Index: 12,08649

Order Now

Specification Test Results

Assay 99.995%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR