2,3,5-Trimethylhydroquinone ≥98%

Supplier: BeanTown Chemical

Synonyms: 2,3,5-Trimethylbenzene-1,4-diol, Trimethylhydroquinone

145015-100G 145015-500G
BT145015-100GEA 50 USD
BT145015-100G BT145015-500G
2,3,5-Trimethylhydroquinone ≥98%
2,3,5-Trimethylhydroquinone

CAS: 700-13-0; EC No: 211-838-3; MDL No: MFCD00002346; RTECS: MX7820000 UN No: UN3077; Haz Class: 9; Packing Group: III Powder; Linear Formula: C6H(CH3)3(OH)2; Molecular Formula: C9H12O2; MW: 152.20 Melting Point: 169-172°; Flash point: 140°C (284°F)


  • RTECS: MX7820000
  • Powder

Formula: C₉H₁₂O₂
MW: 152.19 g/mol
Melting Pt: 169…172 °C
Flash Pt: 140 °C (284 °F)
MDL Number: MFCD00002346
CAS Number: 700-13-0
EINECS: 211-838-3
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR