Sodium tetraphenylborate ≥99.5% ACS
Supplier: BeanTown Chemical
Synonyms:
Tetraphenyl sodium borate, Sodium tetraphenylboron, Tetraphenylboron sodium salt
144880-25G
144880-5G
BT144880-25GEA
151.07
USD
BT144880-25G
BT144880-5G
Sodium tetraphenylborate ≥99.5% ACS
Sodium tetraphenylborate
CAS: 143-66-8; EC No: 205-605-5; MDL No: MFCD00011494; RTECS: ED3362500
UN No: UN2811; Haz Class: 6.1; Packing Group: III
Solid; Linear Formula: (C6H5)4BNa; Molecular Formula: C24H20BNa ; MW: 342.23
Melting Point:
- RTECS: ED3362500
- Light Sensitive
- Solid
Formula:
(C₆H₅)₄BNa MW: 342.22 g/mol Melting Pt: >300 °C |
MDL Number:
MFCD00011494 CAS Number: 143-66-8 EINECS: 205-605-5 UN: 2811 ADR: 6.1,III |
Specification Test Results
Assay | 99.5% min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...