Tris(2-aminoethyl)amine ≥97%

Supplier: BeanTown Chemical

144780-100G 144780-25G 144780-500G
BT144780-100GEA 195.09 USD
BT144780-100G BT144780-25G BT144780-500G
Tris(2-aminoethyl)amine ≥97%
Tris(2-aminoethyl)amine

CAS: 4097-89-6; EC No: 223-857-4; MDL No: MFCD00008177; RTECS: KH8587082 UN No: UN2922; Haz Class: 8 (6.1); Packing Group: II Liquid; Linear Formula: (NH2CH2CH2)3N; Molecular Formula: C6H18N4; MW: 146.24 Boiling Point: 114°/15 mmHg; Flash point: 110°C (230°F) Density (g/mL): 0.976; Refractive Index: 1.497 Air Sensitive , Hygroscopic


  • RTECS: KH8587082
  • Refractive index: 1.497
  • Air Sensitive, Hygroscopic
  • Liquid

Formula: C₆H₁₈N₄
Boiling Pt: 114 °C/15 mmHg
Density: 0.976 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00008177
CAS Number: 4097-89-6
EINECS: 223-857-4
ADR: 8 (6.1),II

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR