1-(3-(Trifluoromethyl)phenyl)piperazine ≥98%
Supplier: BeanTown Chemical
Synonyms:
3-(Trifluoromethyl)-N-phenylpiperazine
144765-25G
144765-5G
BT144765-25GEA
149
USD
BT144765-25G
BT144765-5G
1-(3-(Trifluoromethyl)phenyl)piperazine ≥98%
1-(3-(Trifluoromethyl)phenyl)piperazine
CAS: 15532-75-9; EC No: 239-574-4; MDL No: MFCD00005959; RTECS: TM2820000
UN No: UN2735; Haz Class: 8; Packing Group: III
Liquid; Molecular Formula: C11H13F3N2; MW: 230.23
Flash point: 110°C (230°F)
Density (g/mL): 1.226; Refractive Index: 1.521
Air Sensitive
- RTECS: TM2820000
- Refractive index: 1.521
- Air Sensitive
- Liquid
Formula:
C₁₁H₁₃F₃N₂ Density: 1.226 g/mL Flash Pt: 110 °C (230 °F) |
MDL Number:
MFCD00005959 CAS Number: 15532-75-9 EINECS: 239-574-4 ADR: 8,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...