Crystal violet

Supplier: BeanTown Chemical

Synonyms: CI 42555

144750-100G 144750-25G 144750-500G
BT144750-100GEA 70.22 USD
BT144750-100G BT144750-25G BT144750-500G
Crystal violet
Crystal violet

CAS: 548-62-9; EC No: 208-953-6; MDL No: MFCD00011750; RTECS: BO9000000 UN No: UN3077; Haz Class: 9; Packing Group: III Crystalline/Powder; Molecular Formula: C25H30ClN3; MW: 407.99 Melting Point: 205° (decomposes)


  • RTECS: BO9000000
  • Crystalline/Powder

Formula: C₂₅H₃₀ClN₃
MW: 407.99 g/mol
Melting Pt: 205 °C (dec.)
MDL Number: MFCD00011750
CAS Number: 548-62-9
EINECS: 208-953-6
UN: 3077
ADR: 9 III
Merck Index: 13,04406

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR