Lead(II) oxalate ≥99.9% (trace metals basis)
Supplier: BeanTown Chemical
144745-25G
144745-5G
BT144745-25GEA
153
USD
BT144745-25G
BT144745-5G
Lead(II) oxalate ≥99.9% (trace metals basis)
Lead(II) oxalate
CAS: 814-93-7; EC No: 212-413-5; MDL No: MFCD00040677
UN No: UN2291; Haz Class: 6.1; Packing Group: III
Powder; Linear Formula: PbC2O4; MW: 295.22
- Powder
Formula:
C₂O₄Pb MW: 295.218 g/mol |
MDL Number:
MFCD00040677 CAS Number: 814-93-7 EINECS: 212-413-5 ADR: 6.1 III |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...