Zinc bromide ≥99.9% (trace metals basis)

Supplier: BeanTown Chemical

Synonyms: Zinc (II) bromide

144715-250G 144715-50G
BT144715-1KGEA 577.52 USD
BT144715-1KG BT144715-250G BT144715-50G
Zinc bromide ≥99.9% (trace metals basis)
Zinc bromide

CAS: 7699-45-8; EC No: 231-718-4; MDL No: MFCD00011294; RTECS: ZH1150000 UN No: UN3260; Haz Class: 8; Packing Group: II Crystalline; Linear Formula: ZnBr2; MW: 225.19 Melting Point: 394°; Boiling Point: 697° Density (g/mL): 4.2; Refractive Index: 1.545 Moisture sensitive


  • RTECS: ZH1150000
  • Refractive index: 1.545
  • Moisture sensitive
  • Crystalline

Formula: Br₂Zn
MW: 225.2 g/mol
Boiling Pt: 697 °C
Melting Pt: 394
Density: 4.2 g/mL
MDL Number: MFCD00011294
CAS Number: 7699-45-8
EINECS: 231-718-4
UN: 3260
ADR: 8 II
Merck Index: 13,10182

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR