2,2,6-Trimethylcyclohexanone ≥97%

Supplier: BeanTown Chemical

144665-1G 144665-25G 144665-5G
BT144665-1GEA 55 USD
BT144665-1G BT144665-25G BT144665-5G
2,2,6-Trimethylcyclohexanone ≥97%
2,2,6-Trimethylcyclohexanone

CAS: 2408-37-9; EC No: 219-309-9; MDL No: MFCD00045550 UN No: UN1224; Haz Class: 3; Packing Group: III Liquid; Molecular Formula: C9H16O; MW: 140.23 Flash point: 51°C (125°F) Density (g/mL): 0.904


  • Liquid

Formula: C₉H₁₆O
MW: 140.23 g/mol
Density: 0.904 g/mL
Flash Pt: 51 °C (125 °F)
MDL Number: MFCD00045550
CAS Number: 2408-37-9
EINECS: 219-309-9
UN: 1224
ADR: 3,III

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR