Acetic acid-[D₄] ≥99.5% (Isotopic)
Supplier: BeanTown Chemical
CAS: 1186-52-3; EC No: 214-693-4; MDL No: MFCD00051051
UN No: UN2789; Haz Class: 8 (3); Packing Group: II
Liquid; Linear Formula: CD3CO2D; MW: 64.08
Melting Point: 15-16°; Boiling Point: 115.5°; Flash point: 40°C (104°F)
Density (g/mL): 1.119; Refractive Index: 1.368
Moisture Sensitive
- Refractive index: 1.368
- Moisture Sensitive
- Liquid
Formula:
D₃CCOOD Boiling Pt: 115.5 °C Melting Pt: 15…16 °C Density: 1.119 g/mL Flash Pt: 40 °C (104 °F) |
MDL Number:
MFCD00051051 CAS Number: 1186-52-3 EINECS: 214-693-4 UN: 2789 ADR: 8,II |
Specification Test Results
Assay | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...