Cesium hydroxide hydrate ≥99.9%
Supplier: BeanTown Chemical
Synonyms:
Cesium hydroxide, hydrate
144635-100G
144635-25G
144635-5G
BT144635-100GEA
303.14
USD
BT144635-100G
BT144635-25G
BT144635-5G
Cesium hydroxide hydrate ≥99.9%
Cesium hydroxide hydrate
CAS: 12260-45-6; EC No: 244-344-1; MDL No: MFCD00010964
UN No: UN2682; Haz Class: 8; Packing Group: II
Crystalline; Linear Formula: CsOH·xH2O ; MW: 149.91 (anhydrous)
Melting Point: 272°
Air Sensitive , Hygroscopic
- Air Sensitive, Hygroscopic
- Crystalline
Formula:
HCsO·H₂O MW: 167.93 g/mol Melting Pt: 272 °C |
MDL Number:
MFCD00010964 CAS Number: 12260-45-6 EINECS: 244-344-1 UN: 2682 ADR: 8 II |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...