Copper ≥99.9% (trace metals basis), nanopowder APS 20-50 nm
Supplier: BeanTown Chemical
CAS: 7440-50-8; EC No: 231-159-6; MDL No: MFCD00010965; RTECS: GL5325000
UN No: UN3089; Haz Class: 4.1; Packing Group: II
Powder, CuO layer approx. 2nm; Molecular Formula: Cu; MW: 63.55
Melting Point: 1083.4°; Boiling Point: 2567°
Density (g/mL): 8.94
- RTECS: GL5325000
- Powder, CuO layer approx. 2nm
Formula:
Cu Boiling Pt: 2567 °C Melting Pt: 1083.4 °C Density: 8.94 g/mL |
MDL Number:
MFCD00010965 CAS Number: 7440-50-8 EINECS: 231-159-6 UN: 3089 ADR: 4.1,II |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...