Hexadecylamine ≥90% remainder mainly 1-octadecylamine, Technical Grade

Supplier: BeanTown Chemical

Synonyms: 1-Hexadecylamine, n-Hexadecylamine, 1-Aminohexadecane

144620-1KG 144620-250G
BT144620-1KGEA 111 USD
BT144620-1KG BT144620-250G
Hexadecylamine ≥90% remainder mainly 1-octadecylamine, Technical Grade
Hexadecylamine

CAS: 143-27-1; EC No: 205-596-8; MDL No: MFCD00008158; RTECS: ML9100000 UN No: UN3259; Haz Class: 8; Packing Group: III Crystalline; Linear Formula: CH3(CH2)15NH2; Molecular Formula: C16H35N; MW: 241.46 Melting Point: 43-45°; Boiling Point: 330°; Flash point: 140°C (284°F) Density (g/mL): 0.813


  • RTECS: ML9100000
  • Crystalline

Formula: H₃C(CH₂)₁₄CH₂NH₂
MW: 241.46 g/mol
Boiling Pt: 330 °C
Melting Pt: 43…45 °C
Density: 0.813 g/mL
Flash Pt: 140 °C (284 °F)
MDL Number: MFCD00008158
CAS Number: 143-27-1
EINECS: 205-596-8
UN: 3259
ADR: 8,III

Order Now

Specification Test Results

Assay 90%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR