Bismuth(III) fluoride, anhydrous ≥99%

Supplier: BeanTown Chemical

144525-100G 144525-25G 144525-500G
BT144525-100GEA 121 USD
BT144525-100G BT144525-25G BT144525-500G
Bismuth(III) fluoride, anhydrous ≥99%
Bismuth(III) fluoride

CAS: 7787-61-3; EC No: 232-124-8; MDL No: MFCD00010893 UN No: UN3260; Haz Class: 8; Packing Group: III Powder; Molecular Formula: BiF3; MW: 265.98 Hygroscopic


  • Hygroscopic
  • Powder

Formula: BiF₃
MW: 265.98
Boiling Pt: 230
Density: 3.7
MDL Number: MFCD00010893
CAS Number: 7787-61-3
EINECS: 232-124-8
UN: 3260
ADR: 8 III
Merck Index: 12,01305

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR