Digallium triselenide ≥99.99%

Supplier: BeanTown Chemical

Synonyms: Gallium (III) selenide

144505-1G 144505-5G
BT144505-1GEA 35 USD
BT144505-1G BT144505-5G
Digallium triselenide ≥99.99%
Digallium triselenide

CAS: 12024-24-7; MDL No: MFCD00016103 UN No: UN3077; Haz Class: 9; Packing Group: III 3-12mm Sintered Lump; Molecular Formula: Ga2Se3; MW: 376.33


  • 3-12mm Sintered Lump

Formula: Ga₂Se₃
MW: 376.33 g/mol
MDL Number: MFCD00016103
CAS Number: 12024-24-7
ADR: 9 III

Order Now

Specification Test Results

Assay 99.99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR