α,α,α-Trifluoroacetophenone ≥98%

Supplier: BeanTown Chemical

144490-100G 144490-25G 144490-5G
BT144490-100GEA 298 USD
BT144490-100G BT144490-25G BT144490-5G
α,α,α-Trifluoroacetophenone ≥98%
α,α,α-Trifluoroacetophenone

CAS: 434-45-7; EC No: 207-103-1; MDL No: MFCD00000420 UN No: UN1224; Haz Class: 3; Packing Group: III Liquid; Molecular Formula: C8H5F3O; MW: 174.12 Boiling Point: 165-166°; Flash point: 41°C (106°F) Density (g/mL): 1.24; Refractive Index: 1.458


  • Refractive index: 1.458
  • Liquid

Formula: C₈H₅F₃O
Boiling Pt: 165…166 °C
Density: 1.24 g/mL
Flash Pt: 41 °C (106 °F)
MDL Number: MFCD00000420
CAS Number: 434-45-7
EINECS: 207-103-1
UN: 1224
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=69025716b957e6a38ed2ae6031ab9b0f10246bd1&vid=60781a8070ecc9020bf7794534c2b3fe9acf3ebf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR