Trimethyl orthobutyrate ≥97%
Supplier: BeanTown Chemical
CAS: 43083-12-1; EC No: 256-079-9; MDL No: MFCD00008480
UN No: UN3272; Haz Class: 3; Packing Group: III
Liquid; Linear Formula: CH3CH2CH2C(OCH3)3; Molecular Formula: C7H16O3; MW: 148.20
Boiling Point: 145-157°; Flash point: 50°C (122°F)
Density (g/mL): 0.926; Refractive Index: 1.404
Moisture Sensitive
- Refractive index: 1.404
- Moisture Sensitive
- Liquid
Formula:
C₇H₁₆O₃ Boiling Pt: 145…157 °C Density: 0.926 g/mL Flash Pt: 50 °C (122 °F) |
MDL Number:
MFCD00008480 CAS Number: 43083-12-1 EINECS: 256-079-9 ADR: 3,III |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...