Arsenic ≥99% (trace metals basis), powder -20 mesh

Supplier: BeanTown Chemical

144435-250G 144435-50G
BT144435-250GEA 445 USD
BT144435-250G BT144435-50G
Arsenic ≥99% (trace metals basis), powder -20 mesh
Arsenic

CAS: 7440-38-2; EC No: 213-148-6; MDL No: MFCD00085309; RTECS: CG0525000 UN No: UN1558; Haz Class: 6.1; Packing Group: II Powder; Molecular Formula: As; MW: 74.92 Melting Point: 817°; Boiling Point: 613° (sublimes) Density (g/mL): 5.727


  • RTECS: CG0525000
  • Powder

Formula: As
Boiling Pt: 613 °C (subl.)
Melting Pt: 817 °C
Density: 5.727 g/mL
MDL Number: MFCD00085309
CAS Number: 7440-38-2
EINECS: 213-148-6
UN: 1558
ADR: 6.1,II

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR