Triphenylphosphonium bromide ≥97%
Supplier: BeanTown Chemical
Synonyms:
Triphenylphosphine hydrobromide
144305-25G
144305-5G
BT144305-25GEA
74
USD
BT144305-25G
BT144305-5G
Triphenylphosphonium bromide ≥97%
Triphenylphosphonium bromide
CAS: 6399-81-1; EC No: 229-012-6; MDL No: MFCD00035107
UN No: UN3261; Haz Class: 8; Packing Group: III
Powder; Linear Formula: (C6H5)3P·HBr; Molecular Formula: C18H15P·HBr; MW: 343.21
Melting Point: 196° (decomposes)
Air Sensitive
- Air Sensitive
- Powder
Formula:
C₁₈H₁₆BrP Melting Pt: 196 °C (dec.) |
MDL Number:
MFCD00035107 CAS Number: 6399-81-1 EINECS: 229-012-6 UN: 3261 ADR: 8,III |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...