trans-2-Hexen-1-ol ≥97%

Supplier: BeanTown Chemical

144300-100G 144300-25G 144300-500G
BT144300-100GEA 82 USD
BT144300-100G BT144300-25G BT144300-500G
trans-2-Hexen-1-ol ≥97%
trans-2-Hexen-1-ol

CAS: 928-95-0; EC No: 213-191-2; MDL No: MFCD00002927; RTECS: MP8390000 UN No: UN1987; Haz Class: 3; Packing Group: III Liquid; Linear Formula: CH3CH2CH2CH=CHCH2OH; Molecular Formula: C6H12O; MW: 100.16 Boiling Point: 158-160°; Flash point: 54°C (129°F) Density (g/mL): 0.849; Refractive Index: 1.438


  • RTECS: MP8390000
  • Refractive index: 1.438
  • Liquid

Formula: C₆H₁₂O
Boiling Pt: 158…160 °C
Density: 0.849 g/mL
Flash Pt: 54 °C (129 °F)
MDL Number: MFCD00002927
CAS Number: 928-95-0
EINECS: 213-191-2
UN: 1987
ADR: 3,III

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR