tert-Butanol, anhydrous ≥99.5%

Supplier: BeanTown Chemical

Synonyms: Trimethylcarbinol, tert-Butyl alcohol, 2-Methyl-2-propanol

144290-1L 144290-250ML 144290-4L
BT144290-1LEA 145.96 USD
BT144290-1L BT144290-250ML BT144290-4L
tert-Butanol, anhydrous ≥99.5%
tert-Butanol

CAS: 75-65-0; EC No: 200-889-7; MDL No: MFCD00004464; RTECS: EO1925000 UN No: UN1120; Haz Class: 3; Packing Group: II Liquid; Linear Formula: (CH3)3COH; Molecular Formula: C4H10O ; MW: 74.12 Melting Point: 23-26°; Boiling Point: 83°; Flash point: 11°C (52°F) Density (g/mL): 0.775; Refractive Index: 1.387


  • RTECS: EO1925000
  • Refractive index: 1.387
  • Liquid

Formula: (CH₃)₃COH
Boiling Pt: 83 °C
Melting Pt: 23…26 °C
Density: 0.775 g/mL
Flash Pt: 11 °C (52 °F)
MDL Number: MFCD00004464
CAS Number: 75-65-0
EINECS: 200-889-7
UN: 1120
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=63a1b0c51eec643e5d5b7152a3e410db93d1626e&vid=999328a3c31895b190d09313b24501682921af01&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR