Magnesium oxalate dihydrate ≥98.5%

Supplier: BeanTown Chemical

144225-100G 144225-2KG 144225-500G
BT144225-100GEA 57.08 USD
BT144225-100G BT144225-2KG BT144225-500G
Magnesium oxalate dihydrate ≥98.5%
Magnesium oxalate dihydrate

CAS: 547-66-0; EC No: 208-932-1; MDL No: MFCD00287279 UN No: UN3288; Haz Class: 6.1; Packing Group: III Pieces/Powder; Linear Formula: MgC2O4·2H2O ; MW: 148.36 Melting Point: 150° (decomposes)


  • Pieces/Powder

Formula: C₂MgO₄·2H₂O
MW: 148.36 g/mol (112.32 anhy)
Melting Pt: 150 °C (dec.)
MDL Number: MFCD00287279
CAS Number: 547-66-0
EINECS: 208-932-1
ADR: 6.1 III

Order Now

Specification Test Results

Assay 98.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR