α,α,α',α'-Tetrabromo-o-xylene ≥97%
Supplier: BeanTown Chemical
144220-250G
144220-50G
BT144220-250GEA
245
USD
BT144220-250G
BT144220-50G
α,α,α',α'-Tetrabromo-o-xylene ≥97%
α,α,α',α'-Tetrabromo-o-xylene
CAS: 13209-15-9; EC No: 236-176-2; MDL No: MFCD00000131
UN No: UN1759; Haz Class: 8; Packing Group: II
Crystalline; Linear Formula: C6H4(CHBr2)2; Molecular Formula: C8H6Br4; MW: 421.75
Melting Point: 114-116°
- Crystalline
Formula:
C₈H₆Br₄ Melting Pt: 114…116 °C |
MDL Number:
MFCD00000131 CAS Number: 13209-15-9 EINECS: 236-176-2 UN: 1759 ADR: 6.1,II |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...