Chromium(III) chloride hexahydrate ≥99.5%
Supplier: BeanTown Chemical
Synonyms:
Chromium trichloride hexahydrate, Chromic chloride hexahydrate
144195-100G
144195-500G
BT144195-100GEA
68.17
USD
BT144195-100G
BT144195-500G
Chromium(III) chloride hexahydrate ≥99.5%
Chromium(III) chloride hexahydrate
CAS: 10060-12-5; EC No: 233-038-3; MDL No: MFCD00149660; RTECS: GB5450000
UN No: UN3260; Haz Class: 8; Packing Group: III
Crystalline; Linear Formula: CrCl3·6H2O; MW: 266.45
Density (g/mL): 1.76
Hygroscopic
- RTECS: GB5450000
- Hygroscopic
- Crystalline
Formula:
CrCl₃·6H₂O MW: 266.45 g/mol Melting Pt: 85 Density: 1.76 g/mL |
MDL Number:
MFCD00149660 CAS Number: 10060-12-5 EINECS: 233-038-3 UN: 3260 ADR: 8 III Merck Index: 13,02242 |
Specification Test Results
Assay | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...