Calcium oxide ≥99.95% (trace metals basis)
Supplier: BeanTown Chemical
Synonyms:
Quicklime, Calcium oxide from marble, caustic, Lime
144190-1KG
144190-250G
144190-50G
BT144190-1KGEA
820
USD
BT144190-1KG
BT144190-250G
BT144190-50G
Calcium oxide ≥99.95% (trace metals basis)
Calcium oxide
CAS: 1305-78-8; EC No: 215-138-9; MDL No: MFCD00010911
UN No: UN1910; Haz Class: 8; Packing Group: III
- Air Sensitive, Moisture Sensitive
- <10 Micron APS Powder
Formula:
CaO MW: 56.08 g/mol Boiling Pt: 2850 °C Density: 3.3 g/mL |
MDL Number:
MFCD00010911 CAS Number: 1305-78-8 EINECS: 215-138-9 ADR: 8,III |
Specification Test Results
Assay | 99.95% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...