4-Aminobenzonitrile ≥98%
Supplier: BeanTown Chemical
144160-10G
144160-250G
144160-50G
BT144160-10GEA
32
USD
BT144160-10G
BT144160-250G
BT144160-50G
4-Aminobenzonitrile ≥98%
4-Aminobenzonitrile
CAS: 873-74-5; EC No: 212-850-1; MDL No: MFCD00007821; RTECS: BX2320000
UN No: UN3439; Haz Class: 6.1; Packing Group: III
Crystalline; Molecular Formula: C7H6N2; MW: 118.14
Melting Point: 83-85°
- RTECS: BX2320000
- Crystalline
Formula:
C₇H₆N₂ Melting Pt: 83…85 °C |
MDL Number:
MFCD00007821 CAS Number: 873-74-5 EINECS: 212-850-1 UN: 3439 ADR: 6.1,III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...