cis-1,4-Dichloro-2-butene ≥95%

Supplier: BeanTown Chemical

144125-100G 144125-25G
BT144125-100GEA 321 USD
BT144125-100G BT144125-25G
cis-1,4-Dichloro-2-butene ≥95%
cis-1,4-Dichloro-2-butene

CAS: 1476-11-5; EC No: 216-021-5; MDL No: MFCD00062950; RTECS: EM4900000 UN No: UN3390; Haz Class: 6.1 (8); Packing Group: I Liquid; Linear Formula: ClCH2CH=CHCH2Cl; Molecular Formula: C4H6Cl2; MW: 125.00 Melting Point: -48°; Boiling Point: 152°; Flash point: 55°C (131°F) Density (g/mL): 1.188; Refractive Index: 1.489 Light Sensitive, Moisture Sensitive


  • RTECS: EM4900000
  • Refractive index: 1.489
  • Light Sensitive, Moisture Sensitive
  • Liquid

Formula: C₄H₆Cl₂
MW: 125 g/mol
Boiling Pt: 152 °C
Melting Pt: –48 °C
Density: 1.188 g/mL
Flash Pt: 55 °C (131 °F)
MDL Number: MFCD00062950
CAS Number: 1476-11-5
EINECS: 216-021-5
UN: 3390
ADR: 6.1,I

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR