6-Nitro-o-toluidine ≥98%

Supplier: BeanTown Chemical

Synonyms: 6-Nitro-2-methylaniline, 2-Amino-3-nitrotoluene, 2-Methyl-6-nitroaniline

144095-100G 144095-25G
BT144095-100GEA 313 USD
BT144095-100G BT144095-25G
6-Nitro-o-toluidine ≥98%
6-Nitro-o-toluidine

CAS: 570-24-1; EC No: 209-329-6; MDL No: MFCD00007744 UN No: UN2660; Haz Class: 6.1; Packing Group: III Crystalline; Molecular Formula: C7H8N2O2; MW: 152.15 Melting Point: 93-96°


  • Crystalline

Formula: C₇H₈N₂O₂
MW: 152.15 g/mol
Melting Pt: 93…96 °C
MDL Number: MFCD00007744
CAS Number: 570-24-1
EINECS: 209-329-6
UN: 2660
ADR: 6.1,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR