4-Fluoro-2-(trifluoromethyl)benzonitrile ≥98%

Supplier: BeanTown Chemical

Synonyms: 2-Cyano-5-fluorobenzotrifluoride

144050-1G 144050-25G 144050-5G
BT144050-1GEA 65 USD
BT144050-1G BT144050-25G BT144050-5G
4-Fluoro-2-(trifluoromethyl)benzonitrile ≥98%
4-Fluoro-2-(trifluoromethyl)benzonitrile

CAS: 194853-86-6; MDL No: MFCD00061283 UN No: UN3439; Haz Class: 6.1; Packing Group: III Solid; Molecular Formula: C8H3F4N ; MW: 189.11 Melting Point: 43-45°; Boiling Point: 194-196° Density (g/mL): 1.35


  • Solid

Formula: C₈H₃F₄N
MW: 189.11 g/mol
Boiling Pt: 194…196 °C
Melting Pt: 43…45 °C
Density: 1.35 g/mL
MDL Number: MFCD00061283
CAS Number: 194853-86-6
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR