2-Amino-6-chloro-3-nitropyridine ≥98%
Supplier: BeanTown Chemical
Synonyms:
6-Chloro-3-nitro-2-pyridylamine, 6-Chloro-3-nitropyridin-2-amine
144010-100G
144010-25G
BT144010-100GEA
234
USD
BT144010-100G
BT144010-25G
2-Amino-6-chloro-3-nitropyridine ≥98%
2-Amino-6-chloro-3-nitropyridine
CAS: 27048-04-0; EC No: 248-188-5; MDL No: MFCD00044350
UN No: UN2811; Haz Class: 6.1; Packing Group: III
Powder; Molecular Formula: C5H4ClN3O2 ; MW: 173.56
- Powder
Formula:
C₅H₄ClN₃O₂ MW: 173.56 g/mol |
MDL Number:
MFCD00044350 CAS Number: 27048-04-0 EINECS: 248-188-5 ADR: 6.1,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...