Malachite green oxalate
Supplier: BeanTown Chemical
143990-100G
143990-25G
BT143990-100GEA
88
USD
BT143990-100G
BT143990-25G
Malachite green oxalate
Malachite green oxalate
CAS: 2437-29-8; EC No: 219-441-7; MDL No: MFCD00011766; RTECS: BQ1190000
UN No: UN2811; Haz Class: 6.1; Packing Group: III
Solid; Molecular Formula: C52H54N4O12; MW: 927.02
Melting Point: 144-150°
- RTECS: BQ1190000
- Solid
Formula:
C₅₂H₅₄N₄O₁₂ Melting Pt: 144…150 °C |
MDL Number:
MFCD00011766 CAS Number: 2437-29-8 EINECS: 219-441-7 UN: 2811 ADR: 6.1,III |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...