Cadmium sulfide ≥99.999% (trace metals basis)

Supplier: BeanTown Chemical

Synonyms: Cadmium (II) sulphide

143965-250G 143965-50G
BT143965-250GEA 891 USD
BT143965-250G BT143965-50G
Cadmium sulfide ≥99.999% (trace metals basis)
Cadmium sulphide

CAS: 1306-23-6; EC No: 215-147-8; MDL No: MFCD00010922; RTECS: EV3150000 UN No: UN3077; Haz Class: 9; Packing Group: III 3-12mm Pieces; Molecular Formula: CdS; MW: 144.48 Melting Point: 980° (sublimes) Density (g/mL): 4.82


  • RTECS: EV3150000
  • 3-12mm Pieces

Formula: CdS
MW: 144.48 g/mol
Melting Pt: 980 °C (subl.)
Density: 4.82 g/mL
MDL Number: MFCD00010922
CAS Number: 1306-23-6
EINECS: 215-147-8
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Assay 99.999%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR