o-Xylene ≥99%
Supplier: BeanTown Chemical
CAS: 95-47-6; EC No: 202-422-2; MDL No: MFCD00008519; RTECS: ZE2450000
UN No: UN1307; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: C6H4(CH3)2; Molecular Formula: C8H10O; MW: 106.17
Melting Point: -26 - -23°; Boiling Point: 143-145°; Flash point: 31°C (89°F)
Density (g/mL): 0.879; Refractive Index: 1.505
- RTECS: ZE2450000
- Refractive index: 1.505
- Liquid
Formula:
C₈H₁₀ Boiling Pt: 143…145 °C Melting Pt: –26 – –23 °C Density: 0.879 g/mL Flash Pt: 31 °C (89 °F) |
MDL Number:
MFCD00008519 CAS Number: 95-47-6 EINECS: 202-422-2 UN: 1307 ADR: 3,II |
|
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...