Diisopropylcarbodiimide ≥99%

Supplier: BeanTown Chemical

Synonyms: N,N'-methanetetraylbis(1-methylethylamine), N,N-Diisopropylcarbodiimide, 1,3-Diisopropylcarbodiimide, N,N'-Diisopropylcarbodiimide, N.N’-Diisopropylcarbodiimide, DIC

143890-10G 143890-50G
BT143890-10GEA 37.49 USD
BT143890-10G BT143890-50G
Diisopropylcarbodiimide ≥99%
Diisopropylcarbodiimide

CAS: 693-13-0; EC No: 211-743-7; MDL No: MFCD00065689; RTECS: FF2175000 UN No: UN2929; Haz Class: 6.1 (3); Packing Group: II Liquid; Linear Formula: (CH3)2CHN=C=NCH(CH3)2; Molecular Formula: C7H14N2; MW: 126.20 Boiling Point: 145-148°; Flash point: 31°C (89°F) Density (g/mL): 0.815; Refractive Index: 1.433 Moisture Sensitive


  • RTECS: FF2175000
  • Refractive index: 1.433
  • Moisture Sensitive
  • Liquid

Formula: C₇H₁₄N₂
MW: 126.2 g/mol
Boiling Pt: 145…148 °C
Density: 0.815 g/mL
Flash Pt: 31 °C (89 °F)
MDL Number: MFCD00065689
CAS Number: 693-13-0
EINECS: 211-743-7
UN: 2929
ADR: 6.1,II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR