Isobutyl acetate ≥98%
Supplier: BeanTown Chemical
Synonyms:
2-Methylpropyl acetate
143870-2.5L
143870-500ML
BT143870-2.5LEA
72
USD
BT143870-2.5L
BT143870-500ML
Isobutyl acetate ≥98%
Isobutyl acetate
CAS: 110-19-0; EC No: 203-745-1; MDL No: MFCD00008932; RTECS: AI4022500
UN No: UN1213; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: CH3CO2CH2CH(CH3)2; Molecular Formula: C6H12O2; MW: 116.16
Melting Point: -99°; Boiling Point: 115-117°; Flash point: 18°C (64°F)
Density (g/mL): 0.867; Refractive Index: 1.389
- RTECS: AI4022500
- Refractive index: 1.389
- Liquid
Formula:
C₆H₁₂O₂ MW: 116.16 g/mol Boiling Pt: 115…117 °C Melting Pt: –99 °C Density: 0.867 g/mL Flash Pt: 18 °C (64 °F) |
MDL Number:
MFCD00008932 CAS Number: 110-19-0 EINECS: 203-745-1 UN: 1213 ADR: 3,II |
Specification Test Results
Assay | 98% |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...