cis-Platin

Supplier: BeanTown Chemical

Synonyms: Platinol, cis-Diaminodichloroplatinum(II), cis-Diamminedichloroplatinum (II), cis-Dichlorodiammine platinum(II), Cisplatin, cis-Diamineplatinum(II) dichloride

143840-1G 143840-250MG 143840-5G
BT143840-1GEA 309.95 USD
BT143840-1G BT143840-250MG BT143840-5G
cis-Platin
cis-Platin

CAS: 15663-27-1; EC No: 239-733-8; MDL No: MFCD00011623; RTECS: TP2450000 UN No: UN3288; Haz Class: 6.1; Packing Group: II Crystalline; Linear Formula: Pt(NH3)2Cl2; Molecular Formula: Cl2H6N2Pt; MW: 300.06 Melting Point: 270° (decomposes)


  • RTECS: TP2450000
  • Crystalline

Formula: H₆Cl₂N₂Pt
MW: 300.05 g/mol
Melting Pt: 270 °C (dec.)
MDL Number: MFCD00011623
CAS Number: 15663-27-1
EINECS: 239-733-8
UN: 3288
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR