2-Bromo-5-fluoroaniline ≥98%
Supplier: BeanTown Chemical
143805-25G
143805-5G
BT143805-25GEA
231
USD
BT143805-25G
BT143805-5G
2-Bromo-5-fluoroaniline ≥98%
2-Bromo-5-fluoroaniline
CAS: 1003-99-2; MDL No: MFCD00070750
UN No: UN2811; Haz Class: 6.1; Packing Group: III
Solid; Molecular Formula: C6H5BrFN ; MW: 190.01
Melting Point: 43-47°; Flash point: 110°C (230°F)
- Solid
Formula:
C₆H₅BrFN MW: 190.01 g/mol Melting Pt: 43…47 °C Flash Pt: 110 °C (230 °F) |
MDL Number:
MFCD00070750 CAS Number: 1003-99-2 UN: 2811 ADR: 6.1,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...