Methacrolein ≥96% stabilized
Supplier: BeanTown Chemical
CAS: 78-85-3; EC No: 201-150-1; MDL No: MFCD00006974; RTECS: OZ2625000
UN No: UN2396; Haz Class: 3 (6.1); Packing Group: II
Liquid; Linear Formula: H2C=C(CH3)CHO; Molecular Formula: C4H6O; MW: 70.09
Melting Point: -81°; Boiling Point: 69-70°; Flash point: -15°C (5°F)
Density (g/mL): 0.847; Refractive Index: 1.416
Air Sensitive, Light Sensitive
- RTECS: OZ2625000
- Refractive index: 1.416
- Air Sensitive, Light Sensitive
- Liquid
Formula:
C₄H₆O Boiling Pt: 69…70 °C Melting Pt: –81 °C Density: 0.847 g/mL Flash Pt: –15 °C (5 °F) |
MDL Number:
MFCD00006974 CAS Number: 78-85-3 EINECS: 201-150-1 UN: 2396 ADR: 3,II |
Specification Test Results
Assay | 96% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...