2-Propanol-[D₈] ≥99% (Isotopic)

Supplier: BeanTown Chemical

Synonyms: IPA-D8, 2-Propanol-[²H₈], 2-Hydroxy propane-D8, Isopropyl alcohol-D8, Isopropanol-D8, Dimethyl carbinol-D8

143720-1G 143720-25G 143720-5G
BT143720-1GEA 52.61 USD
BT143720-1G BT143720-25G BT143720-5G
2-Propanol-[D₈] ≥99% (Isotopic)
2-Propanol-[D₈]

CAS: 22739-76-0; EC No: 245-189-2; MDL No: MFCD00044341 UN No: UN1219; Haz Class: 3; Packing Group: II Liquid; Linear Formula: (CD3)2CDOD; Molecular Formula: C3D8O; MW: 68.14 Boiling Point: 82°; Flash point: 12°C (54°F) Density (g/mL): 0.89; Refractive Index: 1.373


  • Refractive index: 1.373
  • Liquid

Formula: D₃CCD(OD)CD₃
Boiling Pt: 82 °C
Density: 0.89 g/mL
Flash Pt: 12 °C (54 °F)
MDL Number: MFCD00044341
CAS Number: 22739-76-0
EINECS: 245-189-2
UN: 1219
ADR: 3,II

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR