p-Cresol ≥99%
Supplier: BeanTown Chemical
Synonyms:
1-Hydroxy-4-methylbenzene, 4-Methylphenol, 4-Hydroxytoluene
143705-1KG
143705-250G
BT143705-1KGEA
78
USD
BT143705-1KG
BT143705-250G
p-Cresol ≥99%
p-Cresol
CAS: 106-44-5; EC No: 203-398-6; MDL No: MFCD00002376; RTECS: GO6475000
UN No: UN3455; Haz Class: 6.1 (8); Packing Group: II
Crystalline; Linear Formula: CH3C6H4OH; Molecular Formula: C7H8O; MW: 108.14
Melting Point: 32-34°; Boiling Point: 201-202°; Flash point: 85°C (185°F)
Density (g/mL): 1.034
Air Sensitive, Hygroscopic, Light Sensitive
- RTECS: GO6475000
- Air Sensitive, Hygroscopic, Light Sensitive
- Crystalline
Formula:
C₇H₈O MW: 108.14 g/mol Boiling Pt: 201…202 °C Melting Pt: 32…34 °C Density: 1.034 g/mL Flash Pt: 85 °C (185 °F) |
MDL Number:
MFCD00002376 CAS Number: 106-44-5 EINECS: 203-398-6 UN: 3455 ADR: 6.1,II |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...