3-(Trifluoromethyl)picolinic acid ≥97%
Supplier: BeanTown Chemical
Synonyms:
3-(Trifluoromethyl)pyridine-2-carboxylic acid, 3-(Trifluoromethy)pyridine-2-carboxylic acid
143680-1G
143680-250MG
BT143680-1GEA
254
USD
BT143680-1G
BT143680-250MG
3-(Trifluoromethyl)picolinic acid ≥97%
3-(Trifluoromethyl)picolinic acid
CAS: 87407-12-3; MDL No: MFCD07375386
UN No: UN2811; Haz Class: 6.1; Packing Group: III
Solid; Molecular Formula: C7H4F3NO2 ; MW: 191.11
Melting Point: 125-130°
- Solid
Formula:
C₇H₄F₃NO₂ MW: 191.1100 g/mol Melting Pt: 125…130 °C |
MDL Number:
MFCD07375386 CAS Number: 87407-12-3 ADR: 6.1,III |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...