Chlorodimethylvinylsilane ≥97%

Supplier: BeanTown Chemical

Synonyms: Dimethylvinylchlorosilane, Vinyldimethylchlorosilane

143670-100G 143670-25G
BT143670-100GEA 164.88 USD
BT143670-100G BT143670-25G
Chlorodimethylvinylsilane ≥97%
Chlorodimethylvinylsilane

CAS: 1719-58-0; EC No: 217-007-1; MDL No: MFCD00018090 UN No: UN2985; Haz Class: 3 (8); Packing Group: II Liquid; Linear Formula: H2C=CHSi(CH3)2Cl; Molecular Formula: C4H9ClSi; MW: 120.66 Boiling Point: 82-83°; Flash point: -5°C (23°F) Density (g/mL): 0.874; Refractive Index: 1.414 Moisture Sensitive


  • Refractive index: 1.414
  • Moisture Sensitive
  • Liquid

Formula: C₄H₉ClSi
MW: 120.65 g/mol
Boiling Pt: 82…83 °C
Density: 0.874 g/mL
Flash Pt: –5 °C (23 °F)
MDL Number: MFCD00018090
CAS Number: 1719-58-0
EINECS: 217-007-1
UN: 2985
ADR: 3,II

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR