3,4-Xylenol ≥98%
Supplier: BeanTown Chemical
CAS: 95-65-8; EC No: 202-439-5; MDL No: MFCD00002304; RTECS: ZE6300000
UN No: UN2261; Haz Class: 6.1; Packing Group: II
Crystalline; Molecular Formula: C8H10O; MW: 122.16
Melting Point: 65-68°; Boiling Point: 227°; Flash point: 110°C (230°F)
- RTECS: ZE6300000
- Crystalline
Formula:
C₈H₁₀O Boiling Pt: 227 °C Melting Pt: 65…68 °C Flash Pt: 110 °C (230 °F) |
MDL Number:
MFCD00002304 CAS Number: 95-65-8 EINECS: 202-439-5 UN: 2261 ADR: 6.1,II |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...