Sodium-tert-pentyloxide ≥95%

Supplier: BeanTown Chemical

Synonyms: Sodium tert-pentoxide, Sodium tert-pentoxide, 95%, tert-Amyl alcohol sodium salt

143560-100G 143560-25G
BT143560-100GEA 58 USD
BT143560-100G BT143560-25G
Sodium-tert-pentyloxide ≥95%
Sodium-tert-pentyloxide

CAS: 14593-46-5; EC No: 238-369-4; MDL No: MFCD00064245 UN No: UN3206; Haz Class: 4.2 (8); Packing Group: II Solid; Linear Formula: NaOC(CH3)2CH2CH3; Molecular Formula: C5H11NaO; MW: 110.13 Air Sensitive, Moisture Sensitive


  • Air Sensitive, Moisture Sensitive
  • Solid

Formula: C₅H₁₁NaO
MW: 110.132 g/mol
MDL Number: MFCD00064245
CAS Number: 14593-46-5
EINECS: 238-369-4
ADR: 4.2 (8),II

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR