Tin(IV) fluoride ≥99%

Supplier: BeanTown Chemical

143505-1G 143505-5G
BT143505-1GEA 41.55 USD
BT143505-1G BT143505-5G
Tin(IV) fluoride ≥99%
Tin(IV) fluoride

CAS: 7783-62-2; EC No: 232-016-0; MDL No: MFCD00042541 UN No: UN2923; Haz Class: 8 (6.1); Packing Group: II -6 Mesh Crystalline; Linear Formula: SnF4; MW: 194.70 Melting Point: 705° (sublimes) Moisture Sensitive


  • Moisture Sensitive
  • -6 Mesh Crystalline

Formula: SnF₄
Melting Pt: 705 °C (subl.)
MDL Number: MFCD00042541
CAS Number: 7783-62-2
EINECS: 232-016-0
ADR: 8 (6.1),II

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR