Tin(II) fluoride ≥97%

Supplier: BeanTown Chemical

Synonyms: Tin difluoride

143490-100G 143490-500G
BT143490-100GEA 42.57 USD
BT143490-100G BT143490-500G
Tin(II) fluoride ≥97%
Tin(II) fluoride

CAS: 7783-47-3; EC No: 231-999-3; MDL No: MFCD00042540; RTECS: XQ3450000 UN No: UN3260; Haz Class: 8; Packing Group: II Crystalline; Linear Formula: SnF2; MW: 156.71 Melting Point: 215°; Boiling Point: 850° Density (g/mL): 4.57


  • RTECS: XQ3450000
  • Crystalline

Formula: SnF₂
MW: 156.71
Boiling Pt: 850
Melting Pt: 215
Density: 4.57 g/mL
MDL Number: MFCD00042540
CAS Number: 7783-47-3
EINECS: 231-999-3
UN: 3260
ADR: 8 II
Merck Index: 12,08940

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR