Sodium bis(trimethylsilyl)amide 2 M in THF
Supplier: BeanTown Chemical
143480-100ML
143480-500ML
BT143480-100MLEA
81.06
USD
BT143480-100ML
BT143480-500ML
Sodium bis(trimethylsilyl)amide 2 M in THF
Sodium bis(trimethylsilyl)amide
CAS: 1070-89-9; EC No: 213-983-8; MDL No: MFCD00009835
UN No: UN2924; Haz Class: 3(8); Packing Group: II
Liquid; Linear Formula: [(CH3)3Si]2NNa; MW: 183.38
Flash point: -17°C (1°F)
Density (g/mL): 0.904
Air Sensitive, Moisture Sensitive
- Air Sensitive, Moisture Sensitive
- Liquid
Formula:
C₆H₁₈NNaSi₂ Density: 0.904 g/mL Flash Pt: –17 °C (1 °F) |
MDL Number:
MFCD00009835 CAS Number: 1070-89-9 EINECS: 213-983-8 UN: 2924 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...