Cobalt(II) carbonate ≥99%

Supplier: BeanTown Chemical

Synonyms: Cobaltous carbonate, Cobalt carbonate, Cobalt(II) carbonate

143470-100G 143470-500G
BT143470-100GEA 73.93 USD
BT143470-100G BT143470-500G
Cobalt(II) carbonate ≥99%
Cobalt(II) carbonate

CAS: 513-79-1; EC No: 208-169-4; MDL No: MFCD00010940 UN No: UN3077; Haz Class: 9; Packing Group: III Powder; Linear Formula: CoCO3; MW: 118.94 Density (g/mL): 4.13


  • Powder

Formula: CCoO₃
MW: 118.94 g/mol
Density: 4.13 g/mL
MDL Number: MFCD00010940
CAS Number: 513-79-1
EINECS: 208-169-4
ADR: 9 III
Merck Index: 12,02497

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR